Learn More
Abnova™ Human LUZP5 Partial ORF (NP_060230, 177 a.a. - 274 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | NP_060230 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 54892 |
Molecular Weight (g/mol) | 36.52kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-953-126
|
Abnova™
H00054892Q01L |
25 ug |
Each for $769.00
|
|
|||||
89-953-125
|
Abnova™
H00054892Q01S |
10 ug |
Each for $499.00
|
|
|||||
Description
Condensin complexes I and II play essential roles in mitotic chromosome assembly and segregation. Both condensins contain 2 invariant structural maintenance of chromosome (SMC) subunits, SMC2 (MIM 605576) and SMC4 (MIM 605575), but they contain different sets of non-SMC subunits. NCAPG2 is 1 of 3 non-SMC subunits that define condensin II (Ono et al., 2003 [PubMed 14532007]).[supplied by OMIM]
Sequence: TKTGADVCRLWRIHQALYCFDYDLEESGEIKDMLLECFININYIKKEEGRRFLSCLFNWNINFIKMIHGTIKNQLQGLQKSLMVYIAEIYFRAWKKASSpecifications
NP_060230 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CAP-G2/FLJ20311/LUZP5/MTB/hCAP-G2 | |
NCAPG2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
54892 | |
LUZP5 (Human) Recombinant Protein (Q01) | |
TKTGADVCRLWRIHQALYCFDYDLEESGEIKDMLLECFININYIKKEEGRRFLSCLFNWNINFIKMIHGTIKNQLQGLQKSLMVYIAEIYFRAWKKAS | |
RUO | |
NCAPG2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.