Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human LYN (BC059394) Full-length Recombinant Protein with GST-tag expressed in Sf9 cells
Used for Func, SDS-PAGE
Supplier: Abnova™ P4199
Specifications
BC059394 | |
Liquid | |
85kDa | |
1 ug/lane SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ26625/JTK8 | |
LYN | |
Baculovirus | |
GST | |
≥90% by SDS-PAGE |
Functional Study, SDS-PAGE | |
4067 | |
LYN (Human) Recombinant Protein | |
5 ug | |
RUO | |
LYN | |
180nmol phosphate incorporated into Poly(Glu-Tyr) per minute per mg protein at 30°C for 15 minutes using a final concentration of 50μM ATP (0.83 uCi/assay). | |
Recombinant | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction