Learn More
Abnova™ Human MAD2L1BP Full-length ORF (AAH02904, 1 a.a. - 274 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00009587P01L
Description
The protein encoded by this gene was identified as a binding protein of the MAD2 mitotic arrest deficient-like 1 (MAD2/MAD2L1). MAD2 is a key component of the spindle checkpoint that delays the onset of anaphase until all the kinetochores are attached to the spindle. This protein may interact with the spindle checkpoint and coordinate cell cycle events in late mitosis. Alternatively spliced transcript variants encoding distinct isoforms have been observed. [provided by RefSeq]
Sequence: MAAPEAEVLSSAAVPDLEWYEKSEETHASQIELLETSSTQEPLNASEAFCPRDCMVPVVFPGPVSQEGCCQFTCELLKHIMYQRQQLPLPYEQLKHFYRKPSPQAEEMLKKKPRATTEVSSRKCQQALAELESVLSHLEDFFARTLVPRVLILLGGNALSPKEFYELDLSLLAPYSVDQSLSTAACLRRLFRAIFMADAFSELQAPPLMGTVVMAQGHRNCGEDWFRPKLNYRVPSRGHKLTVTLSCGRPSIRTTAWEDYIWFQAPVTFKGFRESpecifications
AAH02904 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
55.88kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CMT2/KIAA0110/MGC11282/RP1-261G23.6 | |
MAD2L1BP | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
9587 | |
MAD2L1BP (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAAPEAEVLSSAAVPDLEWYEKSEETHASQIELLETSSTQEPLNASEAFCPRDCMVPVVFPGPVSQEGCCQFTCELLKHIMYQRQQLPLPYEQLKHFYRKPSPQAEEMLKKKPRATTEVSSRKCQQALAELESVLSHLEDFFARTLVPRVLILLGGNALSPKEFYELDLSLLAPYSVDQSLSTAACLRRLFRAIFMADAFSELQAPPLMGTVVMAQGHRNCGEDWFRPKLNYRVPSRGHKLTVTLSCGRPSIRTTAWEDYIWFQAPVTFKGFRE | |
RUO | |
MAD2L1BP | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.