Learn More
Abnova™ Human MAEA Partial ORF (NP_001017405.1, 292 a.a. - 396 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00010296Q01S
Description
This gene product mediates the attachment of erythroblasts to macrophages. This attachment promotes terminal maturation and enucleation of erythroblasts, presumably by suppressing apoptosis. This protein is an integral membrane protein with the N-terminus on the extracellular side and the C-terminus on the cytoplasmic side of the cell. Two immunologically related isoforms of erythroblast macrophage protein with apparent molecular weights of 33 kD and 36 kD were detected in macrophage membranes; this gene encodes the larger isoform. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene, but the biological validity of some variants has not been determined. [provided by RefSeq]
Sequence: AGLSAIKTPQCYKEDGSSKSPDCPVCSRSLNKLAQPLPMAHCANSRLVCKISGDVMNENNPPMMLPNGYVYGYNSLLSIRQDDKVVCPRTKEVFHFSQAEKVYIMSpecifications
NP_001017405.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.29kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
AGLSAIKTPQCYKEDGSSKSPDCPVCSRSLNKLAQPLPMAHCANSRLVCKISGDVMNENNPPMMLPNGYVYGYNSLLSIRQDDKVVCPRTKEVFHFSQAEKVYIM | |
RUO | |
MAEA | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10296 | |
MAEA (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EMLP/EMP/HLC-10/PIG5 | |
MAEA | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.