Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human MAGEF1 Partial ORF (NP_071432, 198 a.a. - 307 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00064110Q01S

 View more versions of this product

Catalog No. 89-951-487


Add to cart

Description

Description

This intronless gene encodes a member of the MAGE superfamily. It is ubiquitously expressed in normal tissues and in tumor cells. This gene includes a microsatellite repeat in the coding region. [provided by RefSeq]

Sequence: VQPSKYHFLFGYPKRLIMEDFVQQRYLSYRRVPHTNPPEYEFSWGPRSNLEISKMEVLGFVAKLHKKEPQHWPVQYREALADEADRARAKARAEASMRARASARAGIHLW
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VQPSKYHFLFGYPKRLIMEDFVQQRYLSYRRVPHTNPPEYEFSWGPRSNLEISKMEVLGFVAKLHKKEPQHWPVQYREALADEADRARAKARAEASMRARASARAGIHLW
RUO
MAGEF1
Wheat Germ (in vitro)
GST
NP_071432
Liquid
64110
MAGEF1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC19617
MAGEF1
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit