Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human MAN2A1 Partial ORF (NP_002363, 1045 a.a. - 1144 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00004124Q01L
Description
This gene encodes a protein which is a member of family 38 of the glycosyl hydrolases. The protein is located in the Golgi and catalyzes the final hydrolytic step in the asparagine-linked oligosaccharide (N-glycan) maturation pathway. Mutations in the mouse homolog of this gene have been shown to cause a systemic autoimmune disease similar to human systemic lupus erythematosus. [provided by RefSeq]
Sequence: DIHLVNLRTIQSKVGNGHSNEAALILHRKGFDCRFSSKGTGLFCSTTQGKILVQKLLNKFIVESLTPSSLSLMHSPPGTQNISEINLSPMEISTFRIQLRSpecifications
NP_002363 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DIHLVNLRTIQSKVGNGHSNEAALILHRKGFDCRFSSKGTGLFCSTTQGKILVQKLLNKFIVESLTPSSLSLMHSPPGTQNISEINLSPMEISTFRIQLR | |
RUO | |
MAN2A1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4124 | |
MAN2A1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GOLIM7/MANA2/MANII | |
MAN2A1 | |
Recombinant | |
wheat germ expression system |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction