Learn More
Abnova™ Human MANEA Full-length ORF (NP_078917.2, 1 a.a. - 462 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Manufacturer: Abnova™ H00079694P01S
Description
N-glycosylation of proteins is initiated in the endoplasmic reticulum (ER) by the transfer of the preassembled oligosaccharide glucose-3-mannose-9-N-acetylglucosamine-2 from dolichyl pyrophosphate to acceptor sites on the target protein by an oligosaccharyltransferase complex. This core oligosaccharide is sequentially processed by several ER glycosidases and by an endomannosidase (E.C. 3.2.1.130), such as MANEA, in the Golgi. MANEA catalyzes the release of mono-, di-, and triglucosylmannose oligosaccharides by cleaving the alpha-1,2-mannosidic bond that links them to high-mannose glycans (Hamilton et al., 2005 [PubMed 15677381]).[supplied by OMIM]
Sequence: MAKFRRRTCIILALFILFIFSLMMGLKMLRPNTATFGAPFGLDLLPELHQRTIHLGKNFDFQKSDRINSETNTKNLKSVEITMKPSKASELNLDELPPLNNYLHVFYYSWYGNPQFDGKYIHWNHPVLEHWDPRIAKNYPQGRHNPPDDIGSSFYPELGSYSSRDPSVIETHMRQMRSASIGVLALSWYPPDVNDENGEPTDNLVPTILDKAHKYNLKVTFHIEPYSNRDDQNMYKNVKYIIDKYGNHPAFYRYKTKTGNALPMFYVYDSYITKPEKWANLLTTSGSRSIRNSPYDGLFIALLVEEKHKYDILQSGFDGIYTYFATNGFTYGSSHQNWASLKLFCDKYNLIFIPSVGPGYIDTSIRPWNTQNTRNRINGKYYEIGLSAALQTRPSLISITSFNEWHEGTQIEKAVPKRTSNTVYLDYRPHKPGLYLELTRKWSEKYSKERATYALDRQLPVSSpecifications
wheat germ expression system | |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
80.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686D20120/ENDO/FLJ12838/hEndo | |
MANEA | |
Yes |
NP_078917.2 | |
Liquid | |
79694 | |
MANEA (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAKFRRRTCIILALFILFIFSLMMGLKMLRPNTATFGAPFGLDLLPELHQRTIHLGKNFDFQKSDRINSETNTKNLKSVEITMKPSKASELNLDELPPLNNYLHVFYYSWYGNPQFDGKYIHWNHPVLEHWDPRIAKNYPQGRHNPPDDIGSSFYPELGSYSSRDPSVIETHMRQMRSASIGVLALSWYPPDVNDENGEPTDNLVPTILDKAHKYNLKVTFHIEPYSNRDDQNMYKNVKYIIDKYGNHPAFYRYKTKTGNALPMFYVYDSYITKPEKWANLLTTSGSRSIRNSPYDGLFIALLVEEKHKYDILQSGFDGIYTYFATNGFTYGSSHQNWASLKLFCDKYNLIFIPSVGPGYIDTSIRPWNTQNTRNRINGKYYEIGLSAALQTRPSLISITSFNEWHEGTQIEKAVPKRTSNTVYLDYRPHKPGLYLELTRKWSEKYSKERATYALDRQLPVS | |
RUO | |
MANEA | |
Wheat Germ (in vitro) | |
GST |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.