Learn More
Abnova™ Human MAOA Full-length ORF (NP_000231.1, 1 a.a. - 527 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Manufacturer: Abnova™ H00004128P02L
Description
This gene encodes monoamine oxidase A, an enzyme that degrades amine neurotransmitters, such as dopamine, norepinephrine, and serotonin. The protein localizes to the mitochondrial outer membrane. The gene is adjacent to a related gene on the opposite strand of chromosome X. Mutation in this gene results in monoamine oxidase deficiency, or Brunner syndrome. [provided by RefSeq]
Sequence: MENQEKASIAGHMFDVVVIGGGISGLSAAKLLTEYGVSVLVLEARDRVGGRTYTIRNEHVDYVDVGGAYVGPTQNRILRLSKELGIETYKVNVSERLVQYVKGKTYPFRGAFPPVWNPIAYLDYNNLWRTIDNMGKEIPTDAPWEAQHADKWDKMTMKELIDKICWTKTARRFAYLFVNINVTSEPHEVSALWFLWYVKQCGGTTRIFSVTNGGQERKFVGGSGQVSERIMDLLGDQVKLNHPVTHVDQSSDNIIIETLNHEHYECKYVINAIPPTLTAKIHFRPELPAERNQLIQRLPMGAVIKCMMYYKEAFWKKKDYCGCMIIEDEDAPISITLDDTKPDGSLPAIMGFILARKADRLAKLHKEIRKKKICELYAKVLGSQEALHPVHYEEKNWCEEQYSGGCYTAYFPPGIMTQYGRVIRQPVGRIFFAGTETATKWSGYMEGAVEAGERAAREVLNGLGKVTEKDIWVQEPESKDVPAVEITHTFWERNLPSVSGLLKIIGFSTSVTALGFVLYKYKLLPRSSpecifications
wheat germ expression system | |
AP, Array, ELISA, WB-Re | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
86.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
25μg | |
RUO | |
MAOA | |
Yes |
NP_000231.1 | |
Liquid | |
4128 | |
Human MAOA Full-length ORF (NP_000231.1, 1 a.a. - 527 a.a.) Recombinant Protein with GST-tag at N-terminal | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at −80°C. Aliquot to avoid repeated freezing and thawing. | |
MAOA | |
Wheat Germ (in vitro) | |
GST Tag |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.