Learn More
Abnova™ Human MAP2K5 (NP_660143.1, 1 a.a. - 448 a.a.) Full-length Recombinant Protein with GST-tag expressed in baculovirus infected Sf21 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P5586
Catalog No. 89-951-593
Add to cart
Description
Description
The protein encoded by this gene is a dual specificity protein kinase that belongs to the MAP kinase kinase family. This kinase specifically interacts with and activates MAPK7/ERK5. This kinase itself can be phosphorylated and activated by MAP3K3/MEKK3, as well as by atypical protein kinase C isoforms (aPKCs). The signal cascade mediated by this kinase is involved in growth factor stimulated cell proliferation and muscle cell differentiation. Four alternatively spliced transcript variants of this gene encoding distinct isoforms have been described. [provided by RefSeq]
Specifications
Specifications
NP_660143.1
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
77kDa
Baculovirus infected insect cell (Sf21) expression system
51% by SDS-PAGE/CBB staining
5 ug
RUO
MAP2K5
The activity was determined by ELISA. The enzyme was incubated with GST-fused substrate protein, and after stopping kinase reaction by EDTA, the reaction solution was transferred into glutathione- coated plate. Phosphorylation was detected by anti-phospho antibody and HRP-labeled anti-rabbit IgG (or HRP-labeled anti-mouse IgG). Substrate: Erk5 [inactive mutant]. ATP: 100 uM.
Yes
Baculovirus infected insect cell (Sf21) expression system
Liquid
5607
MAP2K5 (Human) Recombinant Protein
Glutathione sepharose chromatography
Loading 1 ug protein in SDS-PAGE
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HsT17454|MAPKK5|MEK5|PRKMK5
MAP2K5
Insect
GST