Learn More
Abnova™ Human MAP4K5 Partial ORF (AAH36013, 361 a.a. - 470 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | AAH36013 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 11183 |
Molecular Weight (g/mol) | 37.73kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-951-685
|
Abnova™
H00011183Q01L |
25 ug |
Each for $769.00
|
|
|||||
89-951-684
|
Abnova™
H00011183Q01S |
10 ug |
Each for $499.00
|
|
|||||
Description
This gene encodes a member of the serine/threonine protein kinase family, that is highly similar to yeast SPS1/STE20 kinase. Yeast SPS1/STE20 functions near the beginning of the MAP kinase signal cascades that is essential for yeast pheromone response. This kinase was shown to activate Jun kinase in mammalian cells, which suggested a role in stress response. Two alternatively spliced transcript variants encoding the same protein have been described for this gene. [provided by RefSeq]
Sequence: SSDPNFMLQWNPFVDGANTGKSTSKRAIPPPLPPKPRISSYPEDNFPDEEKASTIKHCPDSESRAPQILRRQSSPSCGPVAETSSIGNGDGISKLMSENTEGSAQAPQLPSpecifications
AAH36013 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GCKR/KHS/KHS1/MAPKKKK5 | |
MAP4K5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
11183 | |
MAP4K5 (Human) Recombinant Protein (Q01) | |
SSDPNFMLQWNPFVDGANTGKSTSKRAIPPPLPPKPRISSYPEDNFPDEEKASTIKHCPDSESRAPQILRRQSSPSCGPVAETSSIGNGDGISKLMSENTEGSAQAPQLP | |
RUO | |
MAP4K5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.