Learn More
Abnova™ Human MAPK1 (NM_002745, 1 a.a. - 360 a.a.) Full-length Recombinant Protein expressed in Sf9 cells
Used for Func, SDS-PAGE
Manufacturer: Abnova™ P4689
Description
The protein encoded by this gene is a member of the MAP kinase family. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. The activation of this kinase requires its phosphorylation by upstream kinases. Upon activation, this kinase translocates to the nucleus of the stimulated cells, where it phosphorylates nuclear targets. Two alternatively spliced transcript variants encoding the same protein, but differing in the UTRs, have been reported for this gene. [provided by RefSeq]
Specifications
NM_002745 | |
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis | |
Liquid | |
41.762kDa | |
Insect cell (Sf9) expression system | |
2 ug/lane SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing | |
ERK/ERK2/ERT1/MAPK2/P42MAPK/PRKM1/PRKM2/p38/p40/p41/p41mapk | |
MAPK1 | |
Insect | |
None |
0.223 ug/uL | |
Liquid | |
5594 | |
MAPK1 (Human) Recombinant Protein | |
GST affinity chromatography | |
100 ug | |
RUO | |
MAPK1 | |
296pmol/μg x min | |
Yes | |
Insect cell (Sf9) expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.