Learn More
Abnova™ Human MAPK13 (NM_002754) Full-length Recombinant Protein with GST expressed in Sf9 cells
Used for Func, SDS-PAGE
Manufacturer: Abnova™ P4237
Description
The protein encoded by this gene is a member of the MAP kinase family. MAP kinases act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. This kinase is closely related to p38 MAP kinase, both of which can be activated by proinflammatory cytokines and cellular stress. MAP kinase kinases 3, and 6 can phosphorylate and activate this kinase. Transcription factor ATF2, and microtubule dynamics regulator stathmin have been shown to be the substrates of this kinase. [provided by RefSeq]
Specifications
NM_002754 | |
Liquid | |
5603 | |
MAPK13 (Human) Recombinant Protein | |
1.5 ug/lane SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC99536/PRKM13/SAPK4/p38delta | |
MAPK13 | |
Insect | |
GST |
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis | |
Liquid | |
71kDa | |
≥80% by SDS-PAGE | |
5 ug | |
RUO | |
MAPK13 | |
259nmol phosphate incorporated into MBP protein per minute per mg protein at 30°C for 15 minutes using a final concentration of 50μM ATP (0.83 uCi/assay). | |
Yes |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.