Learn More
Abnova™ Human MAPK14 (NM_139012) Full-length Recombinant Protein with GST expressed in Sf9 cells
Used for Func, SDS-PAGE
Supplier: Abnova™ P4239
Description
The protein encoded by this gene is a member of the MAP kinase family. MAP kinases act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. This kinase is activated by various environmental stresses and proinflammatory cytokines. The activation requires its phosphorylation by MAP kinase kinases (MKKs), or its autophosphorylation triggered by the interaction of MAP3K7IP1/TAB1 protein with this kinase. The substrates of this kinase include transcription regulator ATF2, MEF2C, and MAX, cell cycle regulator CDC25B, and tumor suppressor p53, which suggest the roles of this kinase in stress related transcription and cell cycle regulation, as well as in genotoxic stress response. Four alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported. [provided by RefSeq]
Specifications
NM_139012 | |
Liquid | |
67kDa | |
SDS-PAGE Result | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CSBP1/CSBP2/CSPB1/EXIP/Mxi2/PRKM14/PRKM15/RK/SAPK2A/p38/p38ALPHA | |
MAPK14 | |
Baculovirus | |
GST | |
≥90% by SDS-PAGE |
Functional Study, SDS-PAGE | |
1432 | |
MAPK14 (Human) Recombinant Protein | |
5 ug | |
RUO | |
MAPK14 | |
The specific activity of p38alpha was determined to be 77nmol/min/mg as per activity assay protocol. | |
Recombinant | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.