Learn More
Abnova™ Human MAPK8IP2 Full-length ORF (AAH47527.1, 1 a.a. - 824 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00023542P02L
Description
The protein encoded by this gene is closely related to MAPK8IP1/IB1/JIP-1, a scaffold protein that is involved in the c-Jun amino-terminal kinase signaling pathway. This protein is expressed in brain and pancreatic cells. It has been shown to interact with, and regulate the activity of MAPK8/JNK1, and MAP2K7/MKK7 kinases. This protein thus is thought to function as a regulator of signal transduction by protein kinase cascade in brain and pancreatic beta-cells. Alternatively spliced transcript variants encoding distinct isoforms have been reported for this gene. [provided by RefSeq]
Sequence: MADRAEMFSLSTFHSLSPPGCRPPQDISLEEFDDEDLSEITDDCGLGLSYDSDHCEKDSLSLGRSEQPHPICSFQDDFQGFEMIDDNEEEDDEDEEEEEEEEEGDGEGQEGGDPGSEAPAPGPLIPSASVEEPHKHRPTTLRLTTLGAQDSLNNNGGFDLVRPASWQETALCSPAPEALRELPGPLPATDTGPGGAQSPVRPGCDCEGNRPAEPPAPGGTSPSSDPGIEADLRSRSSGGRGGRRSSQELSSPGSDSEDAGGARLGRMISSISETELELSSDGGSSSSGRSSHLTNSIEEASSPASEPEPPREPPRRPAFLPVGPDDTNSEYESGSESEPDLSEDADSPWLLSNLVSRMISEGSSPIRCPGQCLSPAPRPPGEPVSPAGGAAQDSQDPEAAAGPGGVELVDMETLCAPPPPAPAAPRPGPAQPGPCLFLSNPTRDTITPLWAAPGRAARPGRACSAACSKEEDEEDDEEEEDAEDSAGSPGGRGTGPSAPRDASLVYDAVKYTLVVDEHTQLELVSLRRCAGLGHDSEEDSGGEASEEEAGAALLGGGQVSGDTSPDSPDLTFSKKFLNVFVNSTSRSSSTESFGLFSCLVNGEEREQTHRAVFRFIPRHPDELELDVDDPVLVEAEEDDFWFRGFNMRTGERGVFPAFYAHAVPGPAKDLLGSKRSPCWVERFDVQFLGSVEVPCHQGNGILCAAMQKIATARKLTVHLRPPASCDLEISLRGVKLSLSGGGPEFQRCSHFFQMKNISFCGCHPRNSCYFGFITKHPLLSRFACHVFVSQESMRPVAQSVGRAFLEYYQEHLAYACPTEDIYLESpecifications
AAH47527.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
114.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IB2/JIP2/PRKM8IPL | |
MAPK8IP2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
23542 | |
MAPK8IP2 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MADRAEMFSLSTFHSLSPPGCRPPQDISLEEFDDEDLSEITDDCGLGLSYDSDHCEKDSLSLGRSEQPHPICSFQDDFQGFEMIDDNEEEDDEDEEEEEEEEEGDGEGQEGGDPGSEAPAPGPLIPSASVEEPHKHRPTTLRLTTLGAQDSLNNNGGFDLVRPASWQETALCSPAPEALRELPGPLPATDTGPGGAQSPVRPGCDCEGNRPAEPPAPGGTSPSSDPGIEADLRSRSSGGRGGRRSSQELSSPGSDSEDAGGARLGRMISSISETELELSSDGGSSSSGRSSHLTNSIEEASSPASEPEPPREPPRRPAFLPVGPDDTNSEYESGSESEPDLSEDADSPWLLSNLVSRMISEGSSPIRCPGQCLSPAPRPPGEPVSPAGGAAQDSQDPEAAAGPGGVELVDMETLCAPPPPAPAAPRPGPAQPGPCLFLSNPTRDTITPLWAAPGRAARPGRACSAACSKEEDEEDDEEEEDAEDSAGSPGGRGTGPSAPRDASLVYDAVKYTLVVDEHTQLELVSLRRCAGLGHDSEEDSGGEASEEEAGAALLGGGQVSGDTSPDSPDLTFSKKFLNVFVNSTSRSSSTESFGLFSCLVNGEEREQTHRAVFRFIPRHPDELELDVDDPVLVEAEEDDFWFRGFNMRTGERGVFPAFYAHAVPGPAKDLLGSKRSPCWVERFDVQFLGSVEVPCHQGNGILCAAMQKIATARKLTVHLRPPASCDLEISLRGVKLSLSGGGPEFQRCSHFFQMKNISFCGCHPRNSCYFGFITKHPLLSRFACHVFVSQESMRPVAQSVGRAFLEYYQEHLAYACPTEDIYLE | |
RUO | |
MAPK8IP2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.