Learn More
Abnova™ Human MARK2 Full-length ORF (NP_001034558.1, 1 a.a. - 755 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Manufacturer: Abnova™ H00002011P01
Description
This gene encodes a member of the Par-1 family of serine/threonine protein kinases. The protein is an important regulator of cell polarity in epithelial and neuronal cells, and also controls the stability of microtubules through phosphorylation and inactivation of several microtubule-associating proteins. The protein localizes to cell membranes. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MIRGRNSATSADEQPHIGNYRLLKTIGKGNFAKVKLARHILTGKEVAVKIIDKTQLNSSSLQKLFREVRIMKVLNHPNIVKLFEVIETEKTLYLVMEYASGGEVFDYLVAHGRMKEKEARAKFRQIVSAVQYCHQKFIVHRDLKAENLLLDADMNIKIADFGFSNEFTFGNKLDTFCGSPPYAAPELFQGKKYDGPEVDVWSLGVILYTLVSGSLPFDGQNLKELRERVLRGKYRIPFYMSTDCENLLKKFLILNPSKRGTLEQIMKDRWMNVGHEDDELKPYVEPLPDYKDPRRTELMVSMGYTREEIQDSLVGQRYNEVMATYLLLGYKSSELEGDTITLKPRPSADLTNSSAPSPSHKVQRSVSANPKQRRFSDQAAGPAIPTSNSYSKKTQSNNAENKRPEEDRESGRKASSTAKVPASPLPGLERKKTTPTPSTNSVLSTSTNRSRNSPLLERASLGQASIQNGKDSLTMPGSRASTASASAAVSAARPRQHQKSMSASVHPNKASGLPPTESNCEVPRPSTAPQRVPVASPSAHNISSSGGAPDRTNFPRGVSSRSTFHAGQLRQVRDQQNLPYGVTPASPSGHSQGRRGASGSIFSKFTSKFVRRNLSFRFARRNLNEPESKDRVETLRPHVVGSGGNDKEKEEFREAKPRSLRFTWSMKTTSSMEPNEMMREIRKVLDANSCQSELHEKYMLLCMHGTPGHEDFVQWEMEVCKLPRLSLNGVRFKRISGTSMAFKNIASKIANELKLSpecifications
wheat germ expression system | |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
110.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
2 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EMK1/MGC99619/PAR-1/Par1b | |
MARK2 | |
Yes |
NP_001034558.1 | |
Liquid | |
2011 | |
MARK2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MIRGRNSATSADEQPHIGNYRLLKTIGKGNFAKVKLARHILTGKEVAVKIIDKTQLNSSSLQKLFREVRIMKVLNHPNIVKLFEVIETEKTLYLVMEYASGGEVFDYLVAHGRMKEKEARAKFRQIVSAVQYCHQKFIVHRDLKAENLLLDADMNIKIADFGFSNEFTFGNKLDTFCGSPPYAAPELFQGKKYDGPEVDVWSLGVILYTLVSGSLPFDGQNLKELRERVLRGKYRIPFYMSTDCENLLKKFLILNPSKRGTLEQIMKDRWMNVGHEDDELKPYVEPLPDYKDPRRTELMVSMGYTREEIQDSLVGQRYNEVMATYLLLGYKSSELEGDTITLKPRPSADLTNSSAPSPSHKVQRSVSANPKQRRFSDQAAGPAIPTSNSYSKKTQSNNAENKRPEEDRESGRKASSTAKVPASPLPGLERKKTTPTPSTNSVLSTSTNRSRNSPLLERASLGQASIQNGKDSLTMPGSRASTASASAAVSAARPRQHQKSMSASVHPNKASGLPPTESNCEVPRPSTAPQRVPVASPSAHNISSSGGAPDRTNFPRGVSSRSTFHAGQLRQVRDQQNLPYGVTPASPSGHSQGRRGASGSIFSKFTSKFVRRNLSFRFARRNLNEPESKDRVETLRPHVVGSGGNDKEKEEFREAKPRSLRFTWSMKTTSSMEPNEMMREIRKVLDANSCQSELHEKYMLLCMHGTPGHEDFVQWEMEVCKLPRLSLNGVRFKRISGTSMAFKNIASKIANELKL | |
RUO | |
MARK2 | |
Wheat Germ (in vitro) | |
GST |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.