Learn More
Abnova™ Human MBNL2 Full-length ORF (AAI04040.1, 1 a.a. - 373 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Manufacturer: Abnova™ H00010150P01L
Description
This gene encodes a C3H-type zinc finger protein, which is similar to the Drosophila melanogaster muscleblind B protein. Drosophila muscleblind is a gene required for photoreceptor differentiation. Several alternatively spliced transcript variants have been described but the full-length natures of only some have been determined. [provided by RefSeq]
Sequence: MALNVAPVRDTKWLTLEVCRQFQRGTCSRSDEECKFAHPPKSCQVENGRVIACFDSLKGRCSRENCKYLHPPTHLKTQLEINGRNNLIQQKTAAAMLAQQMQFMFPGTPLHPVPTFPVGPAIGTNTAISFAPYLAPVTPGVGLVPTEILPTTPVIVPGSPPVTVPGSTATQKLLRTDKLEVCREFQRGNCARGETDCRFAHPADSTMIDTSDNTVTVCMDYIKGRCMREKCKYFHPPAHLQAKIKAAQHQANQAAVAAQAAAAAATVMAFPPGALHPLPKRQALEKSNGTSAVFNPSVLHYQQALTSAQLQQHAAFIPTGSVLCMTPATSIDNSEIISRNGMECQESALRITKHCYCTYYPVSSSIELPQTACSpecifications
wheat germ expression system | |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
66.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp781H1296/MBLL/MBLL39/MGC120625/MGC120626/MGC120628/PRO2032 | |
MBNL2 | |
Yes |
AAI04040.1 | |
Liquid | |
10150 | |
MBNL2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MALNVAPVRDTKWLTLEVCRQFQRGTCSRSDEECKFAHPPKSCQVENGRVIACFDSLKGRCSRENCKYLHPPTHLKTQLEINGRNNLIQQKTAAAMLAQQMQFMFPGTPLHPVPTFPVGPAIGTNTAISFAPYLAPVTPGVGLVPTEILPTTPVIVPGSPPVTVPGSTATQKLLRTDKLEVCREFQRGNCARGETDCRFAHPADSTMIDTSDNTVTVCMDYIKGRCMREKCKYFHPPAHLQAKIKAAQHQANQAAVAAQAAAAAATVMAFPPGALHPLPKRQALEKSNGTSAVFNPSVLHYQQALTSAQLQQHAAFIPTGSVLCMTPATSIDNSEIISRNGMECQESALRITKHCYCTYYPVSSSIELPQTAC | |
RUO | |
MBNL2 | |
Wheat Germ (in vitro) | |
GST |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.