Learn More
Abnova™ Human MBTPS1 Partial ORF (NP_957720.1, 246 a.a. - 355 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | NP_957720.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 8720 |
Molecular Weight (g/mol) | 37.84kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-951-973
|
Abnova™
H00008720Q01S |
10 ug |
Each for $499.00
|
|
|||||
89-951-974
|
Abnova™
H00008720Q01L |
25 ug |
Each for $769.00
|
|
|||||
Description
The encoded protein has a central role in the regulation of lipid metabolism in cells. It is a sterol-regulated subtilisin-like serine protease that cleaves ER membrane-bound sterol regulatory element-binding proteins (SREBPs), a reaction that initiates the two-step proteolytic process by which transcriptionally active fragments of SREBPs are released from the membrane for translocation to the nucleus. The gene product is an integral membrane ER protein, with the bulk located in the ER lumen. It is synthesized as an inactive preproprotein that is self-activated by an intramolecular cleavage that generates the mature protein. [provided by RefSeq]
Sequence: GLGHGTFVAGVIASMRECQGFAPDAELHIFRVFTNNQVSYTSWFLDAFNYAILKKIDVLNLSIGGPDFMDHPFVDKVWELTANNVIMVSAIGNDGPLYGTLNNPADQMDVSpecifications
NP_957720.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0091/MGC138711/MGC138712/PCSK8/S1P/SKI-1 | |
MBTPS1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
8720 | |
MBTPS1 (Human) Recombinant Protein (Q01) | |
GLGHGTFVAGVIASMRECQGFAPDAELHIFRVFTNNQVSYTSWFLDAFNYAILKKIDVLNLSIGGPDFMDHPFVDKVWELTANNVIMVSAIGNDGPLYGTLNNPADQMDV | |
RUO | |
MBTPS1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.