Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human MCAM Full-length ORF (NP_006491.2) Recombinant Protein without tag
Used for AP, Func, Screening
Manufacturer: Abnova™ H00004162G01
Description
Sequence: MGLPRLVCAFLLAACCCCPRVAGVPGEAEQPAPELVEVEVGSTALLKCGLSQSQGNLSHVDWFSVHKEKRTLIFRVRQGQGQSEPGEYEQRLSLQDRGATLALTQVTPQDERIFLCQGKRPRSQEYRIQLRVYKAPEEPNIQVNPLGIPVNSKEPEEVATCVGRNGYPIPQVIWYKNGRPLKEEKNRVHIQSSQTVESSGLYTLQSILKAQLVKEDKDAQFYCELNYRLPSGNHMKESREVTVPVFYPTEKVWLEVEPVGMLKEGDRVEIRCLADGNPPPHFSISKQNPSTREAEEETTNDNGVLVLEPARKEHSGRYECQGLDLDTMISLLSEPQELLVNYVSDVRVSPAAPERQEGSSLTLTCEAESSQDLEFQWLREETGQVLERGPVLQLHDLKREAGGGYRCVASVPSIPGLNRTQLVNVAIFGPPWMAFKERKVWVKENMVLNLSCEASGHPRPTISWNVNGTASEQDQDPQRVLSTLNVLVTPELLETGVECTASNDLGKNTSILFLELVNLTTLTPDSNTTTGLSTSTASPHTRANSTSTERKLPEPESRGVVIVAVIVCILVLAVLGAVLYFLYKKGKLPCRRSGKQEITLPPSRKSELVVEVKSDKLPEEMGLLQGSSGDKRAPGDQGEKYIDLRHSpecifications
wheat germ expression system with proprietary liposome technology | |
Antibody Production, Compound Screening, Functional Study | |
Liquid | |
71.6kDa | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD146/MUC18 | |
MCAM | |
Yes |
NP_006491.2 | |
Liquid | |
4162 | |
MCAM (Human) Recombinant Protein | |
MGLPRLVCAFLLAACCCCPRVAGVPGEAEQPAPELVEVEVGSTALLKCGLSQSQGNLSHVDWFSVHKEKRTLIFRVRQGQGQSEPGEYEQRLSLQDRGATLALTQVTPQDERIFLCQGKRPRSQEYRIQLRVYKAPEEPNIQVNPLGIPVNSKEPEEVATCVGRNGYPIPQVIWYKNGRPLKEEKNRVHIQSSQTVESSGLYTLQSILKAQLVKEDKDAQFYCELNYRLPSGNHMKESREVTVPVFYPTEKVWLEVEPVGMLKEGDRVEIRCLADGNPPPHFSISKQNPSTREAEEETTNDNGVLVLEPARKEHSGRYECQGLDLDTMISLLSEPQELLVNYVSDVRVSPAAPERQEGSSLTLTCEAESSQDLEFQWLREETGQVLERGPVLQLHDLKREAGGGYRCVASVPSIPGLNRTQLVNVAIFGPPWMAFKERKVWVKENMVLNLSCEASGHPRPTISWNVNGTASEQDQDPQRVLSTLNVLVTPELLETGVECTASNDLGKNTSILFLELVNLTTLTPDSNTTTGLSTSTASPHTRANSTSTERKLPEPESRGVVIVAVIVCILVLAVLGAVLYFLYKKGKLPCRRSGKQEITLPPSRKSELVVEVKSDKLPEEMGLLQGSSGDKRAPGDQGEKYIDLRH | |
RUO | |
MCAM | |
Wheat Germ (in vitro) | |
None |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title