Learn More
Abnova™ Human MCM3AP Partial ORF (AAH13285, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00008888Q01L
Description
The minichromosome maintenance protein 3 (MCM3) is one of the MCM proteins essential for the initiation of DNA replication. The protein encoded by this gene is a MCM3 binding protein. It was reported to have phosphorylation-dependent DNA-primase activity, which was up-regulated in antigen immunization induced germinal center. This protein was demonstrated to be an acetyltransferase that acetylates MCM3 and plays a role in DNA replication. The mutagenesis of a nuclear localization signal of MCM3 affects the binding of this protein with MCM3, suggesting that this protein may also facilitate MCM3 nuclear localization. [provided by RefSeq]
Sequence: VRVARCCEDVCAHLVDLFLVEEIFQTAKETLQELQCFCKYLQRWREAVTARKKLRRQMRAFPAAPCCVDVSDRLRALAPSAECPIAEENLARGLLDLGHASpecifications
AAH13285 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VRVARCCEDVCAHLVDLFLVEEIFQTAKETLQELQCFCKYLQRWREAVTARKKLRRQMRAFPAAPCCVDVSDRLRALAPSAECPIAEENLARGLLDLGHA | |
RUO | |
MCM3AP | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8888 | |
MCM3AP (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ44336/FLJ45306/GANP/KIAA0572/MAP80 | |
MCM3AP | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.