Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human MCM6 Full-length ORF (NP_005906.2, 1 a.a. - 821 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00004175P01S

 View more versions of this product

Catalog No. 89-952-043


Add to cart

Description

Description

The protein encoded by this gene is one of the highly conserved mini-chromosome maintenance proteins (MCM) that are essential for the initiation of eukaryotic genome replication. The hexameric protein complex formed by the MCM proteins is a key component of the pre-replication complex (pre_RC) and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. The MCM complex consisting of this protein and MCM2, 4 and 7 proteins possesses DNA helicase activity, and may act as a DNA unwinding enzyme. The phosphorylation of the complex by CDC2 kinase reduces the helicase activity, suggesting a role in the regulation of DNA replication. [provided by RefSeq]

Sequence: MDLAAAAEPGAGSQHLEVRDEVAEKCQKLFLDFLEEFQSSDGEIKYLQLAEELIRPERNTLVVSFVDLEQFNQQLSTTIQEEFYRVYPYLCRALKTFVKDRKEIPLAKDFYVAFQDLPTRHKIRELTSSRIGLLTRISGQVVRTHPVHPELVSGTFLCLDCQTVIRDVEQQFKYTQPNICRNPVCANRRRFLLDTNKSRFVDFQKVRIQETQAELPRGSIPRSLEVILRAEAVESAQAGDKCDFTGTLIVVPDVSKLSTPGARAETNSRVSGVDGYETEGIRGLRALGVRDLSYRLVFLACCVAPTNPRFGGKELRDEEQTAESIKNQMTVKEWEKVFEMSQDKNLYHNLCTSLFPTIHGNDEVKRGVLLMLFGGVPKTTGEGTSLRGDINVCIVGDPSTAKSQFLKHVEEFSPRAVYTSGKASSAAGLTAAVVRDEESHEFVIEAGALMLADNGVCCIDEFDKMDVRDQVAIHEAMEQQTISITKAGVKATLNARTSILAAANPISGHYDRSKSLKQNINLSAPIMSRFDLFFILVDECNEVTDYAIARRIVDLHSRIEESIDRVYSLDDIRRYLLFARQFKPKISKESEDFIVEQYKHLRQRDGSGVTKSSWRITVRQLESMIRLSEAMARMHCCDEVQPKHVKEAFRLLNKSIIRVETPDVNLDQEEEIQMEVDEGAGGINGHADSPAPVNGINGYNEDINQESAPKASLRLGFSEYCRISNLIVLHLRKVEEEEDESALKRSELVNWYLKEIESEIDSEEELINKKRIIEKVIHRLTHYDHVLIELTQAGLKGSTEGSESYEEDPYLVVNPNYLLED
Specifications

Specifications

NP_005906.2
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
119.3kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MCG40308|Mis5|P105MCM
MCM6
Yes
wheat germ expression system
Liquid
4175
MCM6 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MDLAAAAEPGAGSQHLEVRDEVAEKCQKLFLDFLEEFQSSDGEIKYLQLAEELIRPERNTLVVSFVDLEQFNQQLSTTIQEEFYRVYPYLCRALKTFVKDRKEIPLAKDFYVAFQDLPTRHKIRELTSSRIGLLTRISGQVVRTHPVHPELVSGTFLCLDCQTVIRDVEQQFKYTQPNICRNPVCANRRRFLLDTNKSRFVDFQKVRIQETQAELPRGSIPRSLEVILRAEAVESAQAGDKCDFTGTLIVVPDVSKLSTPGARAETNSRVSGVDGYETEGIRGLRALGVRDLSYRLVFLACCVAPTNPRFGGKELRDEEQTAESIKNQMTVKEWEKVFEMSQDKNLYHNLCTSLFPTIHGNDEVKRGVLLMLFGGVPKTTGEGTSLRGDINVCIVGDPSTAKSQFLKHVEEFSPRAVYTSGKASSAAGLTAAVVRDEESHEFVIEAGALMLADNGVCCIDEFDKMDVRDQVAIHEAMEQQTISITKAGVKATLNARTSILAAANPISGHYDRSKSLKQNINLSAPIMSRFDLFFILVDECNEVTDYAIARRIVDLHSRIEESIDRVYSLDDIRRYLLFARQFKPKISKESEDFIVEQYKHLRQRDGSGVTKSSWRITVRQLESMIRLSEAMARMHCCDEVQPKHVKEAFRLLNKSIIRVETPDVNLDQEEEIQMEVDEGAGGINGHADSPAPVNGINGYNEDINQESAPKASLRLGFSEYCRISNLIVLHLRKVEEEEDESALKRSELVNWYLKEIESEIDSEEELINKKRIIEKVIHRLTHYDHVLIELTQAGLKGSTEGSESYEEDPYLVVNPNYLLED
RUO
MCM6
Wheat Germ (in vitro)
GST
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit