Learn More
Abnova™ Human MDFI Full-length ORF (NP_005577.1, 1 a.a. - 246 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | NP_005577.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 4188 |
Molecular Weight (g/mol) | 51.4kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-952-067
|
Abnova™
H00004188P01L |
25 ug |
Each for $769.00
|
|
|||||
89-952-066
|
Abnova™
H00004188P01S |
10 ug |
Each for $499.00
|
|
|||||
Description
This protein is a transcription factor that negatively regulates other myogenic family proteins. Studies of the mouse homolog, I-mf, show that it interferes with myogenic factor function by masking nuclear localization signals and preventing DNA binding. Knockout mouse studies show defects in the formation of vertebrae and ribs that also involve cartilage formation in these structures. [provided by RefSeq]
Sequence: MYQVSGQRPSGCDAPYGAPSAAPGPAQTLSLLPGLEVVTGSTHPAEAAPEEGSLEEAATPMPQGNGPGIPQGLDSTDLDVPTEAVTCQPQGNPLGCTPLLPNDSGHPSELGGTRRAGNGALGGPKAHRKLQTHPSLASQGSKKSKSSSKSTTSQIPLQAQEDCCVHCILSCLFCEFLTLCNIVLDCATCGSCSSEDSCLCCCCCGSGECADCDLPCDLDCGILDACCESADCLEICMECCGLCFSSSpecifications
NP_005577.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
51.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
MYQVSGQRPSGCDAPYGAPSAAPGPAQTLSLLPGLEVVTGSTHPAEAAPEEGSLEEAATPMPQGNGPGIPQGLDSTDLDVPTEAVTCQPQGNPLGCTPLLPNDSGHPSELGGTRRAGNGALGGPKAHRKLQTHPSLASQGSKKSKSSSKSTTSQIPLQAQEDCCVHCILSCLFCEFLTLCNIVLDCATCGSCSSEDSCLCCCCCGSGECADCDLPCDLDCGILDACCESADCLEICMECCGLCFSS | |
RUO | |
MDFI | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
4188 | |
MDFI (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
I-MF/I-mfa | |
MDFI | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.