Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human MED26 Full-length ORF (NP_004822.2, 1 a.a. - 600 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00009441P01S

 View more versions of this product

Catalog No. 89-940-585


Add to cart

Description

Description

The activation of gene transcription is a multistep process that is triggered by factors that recognize transcriptional enhancer sites in DNA. These factors work with co-activators to direct transcriptional initiation by the RNA polymerase II apparatus. The protein encoded by this gene is a subunit of the CRSP (cofactor required for SP1 activation) complex, which, along with TFIID, is required for efficient activation by SP1. This protein is also a component of other multisubunit complexes e.g. thyroid hormone receptor-(TR-) associated proteins which interact with TR and facilitate TR function on DNA templates in conjunction with initiation factors and cofactors. [provided by RefSeq]

Sequence: MTAAPASPQQIRDRLLQAIDPQSNIRNMVAVLEVISSLEKYPITKEALEETRLGKLINDVRKKTKNEELAKRAKKLLRSWQKLIEPAHQHEAALRGLAGATGSANGGAHNCRPEVGAAGPPRSIHDLKSRNDLQRLPGQRLDRLGSRKRRGDQRDLGHPGPPPKVSKASHDPLVPNSSPLPTNGISGSPESFASSLDGSGHAGPEGSRLERDENDKHSGKIPVNAVRPHTSSPGLGKPPGPCLQPKASVLQQLDRVDETPGPPHPKGPPRCSFSPRNSRHEGSFARQQSLYAPKGSVPSPSPRPQALDATQVPSPLPLAQPSTPPVRRLELLPSAESPVCWLEQPESHQRLAGPGCKAGLSPAEPLLSRAGFSPDSSKADSDAASSGGSDSKKKKRYRPRDYTVNLDGQVAEAGVKPVRLKERKLTFDPMTRQIKPLTQKEPVRADSPVHMEQQSRTELDKQEAKASLQSPFEQTNWKELSRNEIIQSYLSRQSSLLSSSGAQTPGAHHFMSEYLKQEESTRQGARQLHVLVPQSPPTDLPGLTREVTQDDLDRIQASQWPGVNGCQDTQGNWYDWTQCISLDPHGDDGRLNILPYVCLD
Specifications

Specifications

NP_004822.2
Liquid
9441
MED26 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTAAPASPQQIRDRLLQAIDPQSNIRNMVAVLEVISSLEKYPITKEALEETRLGKLINDVRKKTKNEELAKRAKKLLRSWQKLIEPAHQHEAALRGLAGATGSANGGAHNCRPEVGAAGPPRSIHDLKSRNDLQRLPGQRLDRLGSRKRRGDQRDLGHPGPPPKVSKASHDPLVPNSSPLPTNGISGSPESFASSLDGSGHAGPEGSRLERDENDKHSGKIPVNAVRPHTSSPGLGKPPGPCLQPKASVLQQLDRVDETPGPPHPKGPPRCSFSPRNSRHEGSFARQQSLYAPKGSVPSPSPRPQALDATQVPSPLPLAQPSTPPVRRLELLPSAESPVCWLEQPESHQRLAGPGCKAGLSPAEPLLSRAGFSPDSSKADSDAASSGGSDSKKKKRYRPRDYTVNLDGQVAEAGVKPVRLKERKLTFDPMTRQIKPLTQKEPVRADSPVHMEQQSRTELDKQEAKASLQSPFEQTNWKELSRNEIIQSYLSRQSSLLSSSGAQTPGAHHFMSEYLKQEESTRQGARQLHVLVPQSPPTDLPGLTREVTQDDLDRIQASQWPGVNGCQDTQGNWYDWTQCISLDPHGDDGRLNILPYVCLD
RUO
MED26
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
91.8kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CRSP7|CRSP70
MED26
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit