Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human MED9 Full-length ORF (NP_060489.1, 1 a.a. - 146 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00055090P01L
Description
The multiprotein Mediator complex is a coactivator required for activation of RNA polymerase II transcription by DNA bound transcription factors. The protein encoded by this gene is thought to be a subunit of the Mediator complex. This gene is located within the Smith-Magenis syndrome region on chromosome 17. [provided by RefSeq]
Sequence: MASAGVAAGRQAEDVLPPTSDQPLPDTKPLPPPQPPPVPAPQPQQSPAPRPQSPARAREEENYSFLPLVHNIIKCMDKDSPEVHQDLNALKSKFQEMRKLISTMPGIHLSPEQQQQQLQSLREQVRTKNELLQKYKSLCMFEIPKESpecifications
NP_060489.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
42.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ10193/MED25/MGC138234 | |
MED9 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
55090 | |
MED9 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MASAGVAAGRQAEDVLPPTSDQPLPDTKPLPPPQPPPVPAPQPQQSPAPRPQSPARAREEENYSFLPLVHNIIKCMDKDSPEVHQDLNALKSKFQEMRKLISTMPGIHLSPEQQQQQLQSLREQVRTKNELLQKYKSLCMFEIPKE | |
RUO | |
MED9 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction