Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human MEF2B Partial ORF (NP_005910.1, 165 a.a. - 235 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00004207Q01S

 View more versions of this product

Catalog No. 89-952-137


Add to cart

Description

Description

This gene represents numerous read-through transcripts that span geneID:729991 and 100271849. Many read-through transcripts are predicted to be nonsense-mediated decay (NMD) candidates, and are thought to be non-coding. Some transcripts are predicted to be capable of translation reinitation at a downstream AUG, resulting in expression of at least one isoform of myocyte enhancer factor 2B (MEF2B) from this read-through locus. At least one additional MEF2B variant and isoform can be expressed from a downstream promoter, and is annotated on geneID:100271849. [provided by RefSeq]

Sequence: SRPSPFRPAAPKAGPPGLVHPLFSPSHLTSKTPPPLYLPTEGRRSDLPGGLAGPRGGLNTSRSLYSGLQNP
Specifications

Specifications

NP_005910.1
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33.44kDa
12.5% SDS-PAGE Stained with Coomassie Blue
SRPSPFRPAAPKAGPPGLVHPLFSPSHLTSKTPPPLYLPTEGRRSDLPGGLAGPRGGLNTSRSLYSGLQNP
RUO
MEF2B
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
4207
MEF2B (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ32599|FLJ46391|MGC189732|MGC189763|RSRFR2
MEF2B
Yes
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit