Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human MEIS1 Partial ORF (NP_002389, 1 a.a. - 90 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00004211Q01L

 View more versions of this product

Catalog No. 89-952-154


Add to cart

Description

Description

Homeobox genes, of which the most well-characterized category is represented by the HOX genes, play a crucial role in normal development. In addition, several homeoproteins are involved in neoplasia. This gene encodes a homeobox protein belonging to the TALE ('three amino acid loop extension') family of homeodomain-containing proteins. [provided by RefSeq]

Sequence: MAQRYDDLPHYGGMDGVGIPSTMYGDPHAARSMQPVHHLNHGPPLHSHQYPHTAHTNAMAPSMGSSVNDALKRDKDAIYGHPLFPLLALI
Specifications

Specifications

NP_002389
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.64kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MAQRYDDLPHYGGMDGVGIPSTMYGDPHAARSMQPVHHLNHGPPLHSHQYPHTAHTNAMAPSMGSSVNDALKRDKDAIYGHPLFPLLALI
RUO
MEIS1
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
4211
MEIS1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC43380
MEIS1
Yes
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit