Learn More
Abnova™ Human MET (NP_000236.2, 956 a.a. - 1390 a.a.) Partial Recombinant Protein with GST-tag expressed in Baculovirus infected Sf21 cells
Used for Func, SDS-PAGE
Manufacturer: Abnova™ P5728
Description
The proto-oncogene MET product is the hepatocyte growth factor receptor and encodes tyrosine-kinase activity. The primary single chain precursor protein is post-translationally cleaved to produce the alpha and beta subunits, which are disulfide linked to form the mature receptor. Various mutations in the MET gene are associated with papillary renal carcinoma. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Specifications
Baculovirus infected insect cell (Sf21) expression system | |
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis | |
Liquid | |
76kDa | |
Baculovirus infected insect cell (Sf21) expression system | |
97% by SDS-PAGE/CBB staining. | |
5 ug | |
RUO | |
MET | |
The activity was measured by off-chip mobility shift assay. The enzyme was incubated with fluorescence-labeled substrate and Mg (or Mn)/ATP. The phosphorylated and unphosphorylated substrates were separated and detected by LabChip™3000. Substrate : Srctide. ATP: 100 µM. |
|
Yes |
NP_000236.2 | |
Liquid | |
4233 | |
MET (Human) Recombinant Protein | |
Glutathione sepharose chromatography | |
Loading 1 ug protein in SDS-PAGE | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AUTS9/HGFR/RCCP2/c-Met | |
MET | |
Insect | |
GST |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.