Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human MFNG Full-length ORF (NP_002396.2, 1 a.a. - 321 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00004242P01S

 View more versions of this product

Catalog No. 89-952-277


Add to cart

Description

Description

This gene is a member of the fringe gene family which also includes Radical and Lunatic fringe. They all encode evolutionarily conserved secreted proteins that act in the Notch receptor pathway to demarcate boundaries during embryonic development. While their genomic structure is distinct from other glycosyltransferases, fringe proteins have a fucose-specific beta1,3 N-acetylglucosaminyltransferase activity that leads to elongation of O-linked fucose residues on Notch, which alters Notch signaling. [provided by RefSeq]

Sequence: MQCRLPRGLAGALLTLLCMGLLCLRYHLNLSPQRVQGTPELSQPNPGPPKLQLHDVFIAVKTTRAFHRLRLELLLDTWVSRTREQTFVFTDSPDKGLQERLGSHLVVTNCSAEHSHPALSCKMAAEFDTFLASGLRWFCHVDDDNYVNPRALLQLLRAFPLARDVYVGRPSLNRPIHASEPQPHNRTRLVQFWFATGGAGFCINRKLALKMAPWASGSRFMDTSALIRLPDDCTMGYIIECKLGGRLQPSPLFHSHLETLQLLRTAQLPEQVTLSYGVFEGKLNVIKLQGPFSPEEDPSRFRSLHCLLYPDTPWCPQLGAR
Specifications

Specifications

NP_002396.2
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
62.6kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MFNG
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
4242
MFNG (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MQCRLPRGLAGALLTLLCMGLLCLRYHLNLSPQRVQGTPELSQPNPGPPKLQLHDVFIAVKTTRAFHRLRLELLLDTWVSRTREQTFVFTDSPDKGLQERLGSHLVVTNCSAEHSHPALSCKMAAEFDTFLASGLRWFCHVDDDNYVNPRALLQLLRAFPLARDVYVGRPSLNRPIHASEPQPHNRTRLVQFWFATGGAGFCINRKLALKMAPWASGSRFMDTSALIRLPDDCTMGYIIECKLGGRLQPSPLFHSHLETLQLLRTAQLPEQVTLSYGVFEGKLNVIKLQGPFSPEEDPSRFRSLHCLLYPDTPWCPQLGAR
RUO
MFNG
Yes
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit