Learn More
Abnova™ Human MGAT4B Full-length ORF (NP_463459.1, 1 a.a. - 563 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00011282P01S
Description
This gene encodes a key glycosyltransferase that regulates the formation of tri- and multiantennary branching structures in the Golgi apparatus. The encoded protein, in addition to the related isoenzyme A, catalyzes the transfer of N-acetylglucosamine (GlcNAc) from UDP-GlcNAc in a beta-1,4 linkage to the Man-alpha-1,3-Man-beta-1,4-GlcNAc arm of R-Man-alpha-1,6(GlcNAc-beta-1,2-Man-alpha-1,3)Man-beta-1,4-GlcNAc-beta-1,4-GlcNAc-beta-1-Asn. The encoded protein may play a role in regulating the availability of serum glycoproteins, oncogenesis, and differentiation. [provided by RefSeq]
Sequence: MSRVAGTRTDVNELLQRWTPRCVRWHTGGARRVALDRPLVTACLPPAGDVVDVYQREFLALRDRLHAAEQESLKRSKELNLVLDEIKRAVSERQALRDGDGNRTWGRLTEDPRLKPWNGSHRHVLHLPTVFHHLPHLLAKESSLQPAVRVGQGRTGVSVVMGIPSVRREVHSYLTDTLHSLISELSPQEKEDSVIVVLIAETDSQYTSAVTENIKALFPTEIHSGLLEVISPSPHFYPDFSRLRESFGDPKERVRWRTKQNLDYCFLMMYAQSKGIYYVQLEDDIVAKPNYLSTMKNFALQQPSEDWMILEFSQLGFIGKMFKSLDLSLIVEFILMFYRDKPIDWLLDHILWVKVCNPEKDAKHCDRQKANLRIRFKPSLFQHVGTHSSLAGKIQKLKDKDFGKQALRKEHVNPPAEVSTSLKTYQHFTLEKAYLREDFFWAFTPAAGDFIRFRFFQPLRLERFFFRSGNIEHPEDKLFNTSVEVLPFDNPQSDKEALQEGRTATLRYPRSPDGYLQIGSFYKGVAEGEVDPAFGPLEALRLSIQTDSPVWVILSEIFLKKADSpecifications
NP_463459.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
91.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GNT-IV/GNT-IVB | |
MGAT4B | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
11282 | |
MGAT4B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSRVAGTRTDVNELLQRWTPRCVRWHTGGARRVALDRPLVTACLPPAGDVVDVYQREFLALRDRLHAAEQESLKRSKELNLVLDEIKRAVSERQALRDGDGNRTWGRLTEDPRLKPWNGSHRHVLHLPTVFHHLPHLLAKESSLQPAVRVGQGRTGVSVVMGIPSVRREVHSYLTDTLHSLISELSPQEKEDSVIVVLIAETDSQYTSAVTENIKALFPTEIHSGLLEVISPSPHFYPDFSRLRESFGDPKERVRWRTKQNLDYCFLMMYAQSKGIYYVQLEDDIVAKPNYLSTMKNFALQQPSEDWMILEFSQLGFIGKMFKSLDLSLIVEFILMFYRDKPIDWLLDHILWVKVCNPEKDAKHCDRQKANLRIRFKPSLFQHVGTHSSLAGKIQKLKDKDFGKQALRKEHVNPPAEVSTSLKTYQHFTLEKAYLREDFFWAFTPAAGDFIRFRFFQPLRLERFFFRSGNIEHPEDKLFNTSVEVLPFDNPQSDKEALQEGRTATLRYPRSPDGYLQIGSFYKGVAEGEVDPAFGPLEALRLSIQTDSPVWVILSEIFLKKAD | |
RUO | |
MGAT4B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.