Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human MGC42105 (NP_699192.1, 1 a.a. - 436 a.a.) Full-length Recombinant Protein with GST-tag expressed in baculovirus infected Sf21 cells
Used for Func, SDS-PAGE
Manufacturer: Abnova™ P5597
Specifications
Baculovirus infected insect cell (Sf21) expression system | |
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis | |
Liquid | |
76kDa | |
Baculovirus infected insect cell (Sf21) expression system | |
86% by SDS-PAGE/CBB staining | |
5 ug | |
RUO | |
MGC42105 | |
The activity was measured by off-chip mobility shift assay. The enzyme was incubated with fluorescence-labeled substrate and Mg(or Mn)/ATP. The phosphorylated and unphosphorylated substrates were separated and detected by LabChip 3000. Substrate: CHKtide. ATP: 100 uM. | |
Yes |
NP_699192.1 | |
Liquid | |
167359 | |
MGC42105 (Human) Recombinant Protein | |
Glutathione sepharose chromatography and anion exchange chromatography | |
Loading 1 ug protein in SDS-PAGE | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NIM1 | |
MGC42105 | |
Insect | |
GST |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title