Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Mindin Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
$200.50 - $549.50
Specifications
Antigen | Mindin/Spondin-2 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Mindin/Spondin-2 Polyclonal specifically detects Mindin/Spondin-2 in Human samples. It is validated for Western Blot.Specifications
Mindin/Spondin-2 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
Differentially expressed in cancerous and non-cancerous lung cells 1, DIL1, DIL-1, DIL1FLJ34460, DKFZp686G21139, FLJ16313, FLJ22401, MINDIN, M-SPONDIN, RG-1, SPON2, Spondin 2, spondin 2, extracellular matrix protein, spondin-2 | |
SPON2 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q9BUD6 | |
10417 | |
Mouse myeloma cell line NS0-derived recombinant human Mindin Glu32-Val331 Accession # Q9BUD6 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Mindin in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title