Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MIP-1 alpha (aa 27-92), Recombinant Protein, Invitrogen™ Non-distribution product as customer accommodation.

E. coli Recombinant Protein

$115.60 - $2310.00

Specifications

Recombinant Yes
Name Human MIP-1 alpha (aa 27-92)
For Use With (Application) Control, ELISA, Western Blot
Concentration 200 µg/mL
Conjugate Unlabeled
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIRP87849 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87849
10 µg
Each for $115.60
Add to cart
 
PIRP87850 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87850
25 µg
Each for $289.00
Add to cart
 
PIRP87851 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87851
100 µg
Each for $764.00
Add to cart
 
PIRP87852 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87852
500 µg
Each for $2,310.00
Add to cart
 
Description

Description

Macrophage inflammatory protein-1a (MIP-1a) is an 8kDa member of the C-C chemokine subfamily. C-C chemokines are chemoattractants and activators for monocytes and T cells. MIP-1a has been identified as one of the major HIV-SFs produced by CD8+ T cells, along with MIP-1b and RANTES.
Specifications

Specifications

Yes
Control, ELISA, Western Blot
Unlabeled
4 mM HCl with no preservative
E. coli
-20°C
P10147
Ccl3
>95%
Human MIP-1 alpha (aa 27-92)
200 µg/mL
Liquid
AI323804; C-C motif chemokine 3; C-C motif chemokine ligand 3; CCL3; chemokine (C-C motif) ligand 3; G0/G1 switch regulatory protein 19-1; G0S19-1; Heparin-binding chemotaxis protein; H-MIP-1-alpha; L2G25B; LD78alpha; LD78-alpha; LD78-alpha(4-69); Macrophage inflammatory protein 1 alpha (Small inducible cytokine A3); macrophage inflammatory protein 1-alpha; macrophage inflammatory protein-1 alpha; MIP1 (a); mip1 alpha; MIP-1 alpha; MIP1-(a); Mip1a; MIP-1 A; MIP-1 alpha; MIP-1-alpha; MIP1-alpha; MIP-1-alpha(4-69); PAT 464.1; RP23-320E6.7; SCYA3; SIS-alpha; SIS-beta; small inducible cytokine A3; small inducible cytokine A3 (homologous to mouse Mip-1 A); small-inducible cytokine A3; Tonsillar lymphocyte LD78 alpha protein; TY-5
RUO
Human
6348
7.5 kD
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit