Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MIP-3 beta (aa 22-98), Recombinant Protein, Invitrogen™ Non-distribution product as customer accommodation.

E. coli Recombinant Protein

$160.50 - $1959.00

Specifications

Recombinant Yes
Name Human MIP-3 beta (aa 22-98)
For Use With (Application) Control, ELISA, Western Blot
Concentration 200 µg/mL
Conjugate Unlabeled
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIRP87845 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87845
10 µg
Each for $160.50
Add to cart
 
PIRP87846 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87846
25 µg
Each for $276.50
Add to cart
 
PIRP87847 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87847
100 µg
Each for $751.50
Add to cart
 
PIRP87848 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87848
500 µg
Each for $1,959.00
Add to cart
 
Description

Description

CCL19, also known as MIP-3 beta (Macrophage Inflammatory Protein 3 beta), is a member of the CC- subfamily of chemokines. It is most closely related to CCL21, with which it shares 32% amino acid sequence homology. CCL19 and CCL21 are expressed mainly by stromal cells in the T cell-rich zones of lymph nodes. They are critical mediators of the homeostatic trafficking of naive T cells and activated dendritic cells into the secondary lymphoid organs. CCL19 and CCL21 also play a role in T cell priming and activation, as well as the recruitment of lymphocytes to inflamed tissue. Both proteins signal via the G protein-coupled receptor, CCR7, which is expressed on T cells and mature dendritic cells. Although CCL19 and CCL21 both exhibit the same affinity for CCR7, only the binding of CCL19 results in the desensitization and internalization of the receptor.
Specifications

Specifications

Yes
Control, ELISA, Western Blot
Unlabeled
PBS with no preservative
E. coli
-20°C
Q99731
Ccl19
>95%
Human MIP-3 beta (aa 22-98)
200 µg/mL
Liquid
beta chemokine exodus-3; beta-chemokine exodus-3; CC chemokine ligand 19; C-C motif chemokine; C-C motif chemokine 19; C-C motif chemokine ligand 19; Ccl19; chemokine (C-C motif) ligand 19; chemokine CCL19; CK beta-11; CKb11; EBI1 ligand chemokine; EBI-1 ligand chemokine; EBI1-ligand chemokine; EBV-induced molecule 1 ligand chemokine; ELC; Epstein-Barr virus-induced molecule 1 ligand chemokine; exodus-3; Macrophage inflammatory protein 3 beta; macrophage inflammatory protein 3-beta; MIP3B; MIP-3 b; MIP-3-beta; M-MIP-3-beta; Scya19; small inducible cytokine A19; small inducible cytokine subfamily A (Cys-Cys), member 19; small-inducible cytokine A19
RUO
Human
6363
8.8 kD
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit