Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MITF Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF5769SP
This item is not returnable.
View return policy
Description
MITF Polyclonal specifically detects MITF in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MITF | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BHLHE32, bHLHe32MI, Class E basic helix-loop-helix protein 32, MI, microphthalmia-associated transcription factor, Waardenburg syndrome, type 2A, WS2A | |
Goat | |
Affinity Purified | |
RUO | |
4286 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
O75030 | |
MITF | |
E. coli-derived recombinant human MITF Val119-Lys289 Accession # O75030 | |
25 μg | |
Primary | |
Detects human MITF in direct ELISAs and Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction