Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human MKNK2 (NM_017572) Full-length Recombinant Protein with GST-tag expressed in Sf9 cells
Used for Func, SDS-PAGE
Manufacturer: Abnova™ P3882
Specifications
Insect cell (Sf9) expression system | |
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis | |
2872 | |
MKNK2 (Human) Recombinant Protein | |
>70% by densitometry | |
5 ug | |
RUO | |
MKNK2 | |
The specific activity was determined to be 93nmol /min/mg as per Activity Assay Protocol. | |
Yes |
NM_017572 | |
In 50mM Tris-HCl, 150mM NaCl, pH 7.5 (0.25mM DTT, 0.1mM EGTA, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol) | |
41kDa | |
Insect cell (Sf9) expression system | |
SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GPRK7/MNK2 | |
MKNK2 | |
Insect | |
GST |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title