Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MMP-19 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF6790
This item is not returnable.
View return policy
Description
MMP-19 Polyclonal specifically detects MMP-19 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MMP-19 | |
Polyclonal | |
Unconjugated | |
Q99542 | |
MMP19 | |
Mouse myeloma cell line NS0-derived recombinant human MMP-19 Leu229-Tyr508 Accession # Q99542 | |
100 μg | |
Primary | |
Detects human MMP-19 in direct ELISAs. | |
Human | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
EC 3.4.24.-, matrix metallopeptidase 19, matrix metalloproteinase 18, matrix metalloproteinase 19, Matrix metalloproteinase RASI, Matrix metalloproteinase-18, matrix metalloproteinase-19, MMP-18, MMP18MMP-19, MMP19, RASI, RASI-1 | |
Goat | |
Affinity Purified | |
RUO | |
4327 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction