Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Invitrogen™ eBioscience™ Human MMP-3 Platinum ELISA Kit Non-distribution product as customer accommodation.
For the quantitative detection of total human MMP-3.
Manufacturer:  Invitrogen™ BMS20143
 View more versions of this product
Catalog No. 50-112-5245
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human MMP-3
Description
Description
MMP3 (stromelysin-1) belongs to the peptidase M10A subfamily. It is secreted in its pro-protein form, and is activated by extracellular proteinase cleavage. MMP3 is capable of binding to zinc and calcium ions, and has metallo-endopeptidase activity. The active enzyme degrades collagen, gelatin, fibronectin, and laminin. It is involved in normal cellular processes such as wound repair and tissue remodeling. Mutations in the gene can result in coronary heart disease 6.
Specifications
Specifications
P08254
ELISA
Colorimetric Microplate Reader
ELISA Kit
RUO
Plasma,Serum,Supernatant
Human
1 hr 20 min
Human
Matrix metalloproteinase-3, Stromelysin-1, Progelatinase, Transin
6.1%
0.06-4.00 ng/mL
4314
Adhesive Plate Covers
96 tests
0.0005 ng/mL
Plasma,10 μL|Serum,10 μL|Supernatant,10 μL
2-8°C
3 hr 30 min
MMP3
4%
HRP