Learn More
Abnova™ Human MMRN1 Partial ORF (NP_031377, 291 a.a. - 390 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00022915Q01L
Description
Multimerin is a massive, soluble protein found in platelets and in the endothelium of blood vessels. It is comprised of subunits linked by interchain disulfide bonds to form large, variably sized homomultimers. Multimerin is a factor V/Va-binding protein and may function as a carrier protein for platelet factor V. It may also have functions as an extracellular matrix or adhesive protein. Recently, patients with an unusual autosomal-dominant bleeding disorder (factor V Quebec) were found to have a deficiency of platelet multimerin. [provided by RefSeq]
Sequence: IHTNQAESHTAVGRGVAEQQQQQGCGDPEVMQKMTDQVNYQAMKLTLLQKKIDNISLTVNDVRNTYSSLEGKVSEDKSREFQSLLKGLKSKSINVLIRDISpecifications
NP_031377 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
IHTNQAESHTAVGRGVAEQQQQQGCGDPEVMQKMTDQVNYQAMKLTLLQKKIDNISLTVNDVRNTYSSLEGKVSEDKSREFQSLLKGLKSKSINVLIRDI | |
RUO | |
MMRN1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
22915 | |
MMRN1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ECM/EMILIN4/GPIa*/MMRN | |
MMRN1 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.