Learn More
Novus Biologicals™ Human Motilin ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP266719
Catalog No. NBP266719
Add to cart
Description
Description
The Human Motilin ELISA Kit (Colorimetric) measures human Motilin in serum, plasma and other biological fluids.
Specifications
Specifications
15.63 - 1000 pg/mL
This kit recognizes Human MTL in samples. No significant cross-reactivity or interference between Human MTL and analogues was observed.
4295
Human
1 Kit
Storage is content dependent.
9.38 pg/mL
ELISA
MLN
ELISA Kit (Colorimetric)
Biological Fluids,Plasma,Serum
Motilin