Learn More
Novus Biologicals™ Human/Mouse GM-CSF ELISA Kit
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP191277
Catalog No. NBP191277
Add to cart
Description
Description
The Human, Mouse GM-CSF ELISA Kit (Colorimetric) measures human, mouse GM-CSF in cell culture supernatant, serum, plasma (edta, heparin).
Specifications
Specifications
7.8 - 500pg/mL
1437
ELISA Kit
2.9pg/mL
Human, Mouse (Hu, Mu)
GM-CSF
CSF2
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human GM-CSF, HRP-Conjugate anti-human GM-CSF monoclonal antibody, Human GM-CSF Standard lyophilized, 1ng/mL upon reconstitution, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
96 Tests
Cell culture supernatant, serum, plasma (EDTA, heparin)
Storage of components varies. See protocol for specific instructions.
Cytokine Research, Immunology
colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, molgramostin, sargramostim