Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse MMP-24/MT5-MMP Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $655.00
Specifications
Antigen | MMP-24/MT5-MMP |
---|---|
Clone | 143908 |
Dilution | Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, CyTOF |
Classification | Monoclonal |
Description
MMP-24/MT5-MMP Monoclonal specifically detects MMP-24/MT5-MMP in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
MMP-24/MT5-MMP | |
Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 3.4.24, EC 3.4.24.-, EC 3.4.24.80, matrix metallopeptidase 24 (membrane-inserted), matrix metalloproteinase 24 (membrane-inserted), matrix metalloproteinase-24, membrane-type 5 matrix metalloproteinase, Membrane-type matrix metalloproteinase 5, Membrane-type-5 matrix metalloproteinase, MMP24, MMP-24, MT5MMP, MT5-MMP, MT5-MMPMMP25, MT-MMP 5, MTMMP5, MT-MMP5 | |
MMP24 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
143908 | |
Western Blot, Flow Cytometry, CyTOF | |
Unconjugated | |
Rat | |
Human, Mouse | |
Q9R0S2 | |
10893 | |
E. coli-derived recombinant mouse MMP-24/MT5-MMP Arg125-Gln296 Accession # Q9R0S2 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse MMP-24/MT5-MMP in direct ELISAs and detects human and mouse MMP-24/MT5-MMP in Western blots. In Western blots, less than 5% cross-reactivity with recombinant mouse (rm) MMP-3, rmMMP-9, and the catalytic domains of recombinant human MMP-14, 15, and 16 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title