Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Peroxiredoxin 5 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3774
This item is not returnable.
View return policy
Description
Peroxiredoxin 5 Polyclonal specifically detects Peroxiredoxin 5 in Human, Mouse samples. It is validated for Western Blot.Specifications
Peroxiredoxin 5 | |
Polyclonal | |
Unconjugated | |
P30044 | |
PRDX5 | |
E. coli-derived recombinant human Peroxiredoxin 5 Met1-Leu214 Accession # P30044 | |
100 μg | |
Primary | |
Detects endogenous human and mouse Peroxiredoxin 5 in Western blots. In Western blots, this antibody shows no cross-reactivity with recombinant human Peroxiredoxin 1, 2, 3, or 6. | |
Human, Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.5 ug/mL | |
ACR1, ACR1Peroxisomal antioxidant enzyme, Alu corepressor 1, Alu co-repressor 1, Antioxidant enzyme B166, AOEB166, AOEB166Peroxiredoxin V, AOPP, B166, EC 1.11.1.15, Liver tissue 2D-page spot 71B, MGC117264, MGC142283, MGC142285, peroxiredoxin 5, peroxiredoxin-5, mitochondrial, PLPThioredoxin peroxidase PMP20, PMP20, PRDX5, PRDX6, prx-V, PRXV, SBBI10, Thioredoxin reductase, TPx type VI | |
Goat | |
Affinity Purified | |
RUO | |
25824 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction