Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human/Mouse Phospho-Akt1 (S473) DuoSet IC ELISA
Human/Mouse Phospho-Akt1 (S473) DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Specifications
Product Type | ELISA Development Kit for Phosphorylated Intracellular Proteins |
---|---|
Species | Human, Mouse |
Gene ID (Entrez) | 207 |
Catalog Number | Mfr. No. | Packaging Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Packaging Quantity | Price | Quantity & Availability | |||||
DYC2289CE
|
R&D Systems™
DYC2289CE |
15 Plates | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
DYC2289C2
|
R&D Systems™
DYC2289C2 |
2 Plates | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
DYC2289C5
|
R&D Systems™
DYC2289C5 |
5 Plates | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
The Human/Mouse Phospho-Akt1 (S473) DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human, mouse AKT1.Specifications
ELISA Development Kit for Phosphorylated Intracellular Proteins | |
Human, Mouse | |
207 |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title