Learn More
R&D Systems™ Human/Mouse/Rat Activin A Recombinant Protein Available on GSA/VA Contract for Federal Government customers only.
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
$3970.00
Specifications
Product Type Recombinant protein
Regulatory Status RUO
Recombinant Yes
Biological Activity 0.2 to 1.2ng/mL
Quantity 500μg
View More Specs
Products 2
Catalog Number Mfr. No. Formulation Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Price Quantity & Availability  
33-8AC-500 Available on GSA/VA Contract for Federal Government customers only. View Documents R&D Systems™
338AC500
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein Each for $3,970.00
Add to cart
33-8AC-500CF Available on GSA/VA Contract for Federal Government customers only. View Documents R&D Systems™
338AC500/CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA Each for $3,970.00
Add to cart
Description
Description
The Recombinant Human/Mouse/Rat Activin A Protein is derived from CHO. The Recombinant Human/Mouse/Rat Activin A Protein has been validated for the following applications: Bioactivity.
Order Info
Ships at ambient temperature.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Recombinant protein
Yes
500μg
>95%, by SDS-PAGE with silver Staining
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
INHBA
P08476
Bioactivity
RUO
0.2 to 1.2ng/mL
Unlabeled
Human, Mouse, Rat
13 kDa (monomer)
<0.01 EU per 1μg of the protein by the LAL method
Chinese Hamster Ovary cell line, CHO-derived Gly311-Ser426