Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ gp96/HSP90B1/GRP94 ELISA Development Kit

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

Manufacturer:  Novus Biologicals™ NBP262146

Catalog No. NBP262146


Add to cart

Description

Description

Assay has high sensitivity and specificity for detection of  Human, Mouse, Rat, Canine gp96/HSP90B1/GRP94.
Specifications

Specifications

5 Kit
6.25 - 200 ng/ml
ELISA
ELISA Kit (Colorimetric)
Store at 4°C
Human, Mouse, Rat, Canine
Cell Lysates, Microsomes
Cross reactivity with human Calreticulin, PDI, HSP90alpha, HSP90beta, HSP70, hamster Grp78 and bovine Hsc70 is less than 0.035%.
7184
1.29 ng/ml
gp96/HSP90B1/GRP94
HSP90B1
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit