Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat DEP-1/CD148 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Supplier: R&D Systems AF1934
This item is not returnable.
View return policy
Description
DEP-1/CD148 Polyclonal specifically detects DEP-1/CD148 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation.Specifications
DEP-1/CD148 | |
Polyclonal | |
Unconjugated | |
Q12913 | |
PTPRJ | |
E. coli-derived recombinant human DEP-1/CD148 Arg997-Ala1337 Accession # Q12913 | |
100 μg | |
Primary | |
Detects human, mouse, and rat DEP-1/CD148 in Western blots. | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunoprecipitation | |
LYOPH | |
Western Blot 1 ug/mL, Immunoprecipitation 2.5 ug/500 ug cell lysate | |
CD148, CD148 antigen, Density-enhanced phosphatase 1, DEP1, DEP-1, DEP1EC 3.1.3.48, HPTP eta, HPTPeta, HPTP-eta, human density enhanced phosphatase-1, protein tyrosine phosphatase, receptor type, J, protein tyrosine phosphatase, receptor type, J polypeptide, Protein-tyrosine phosphatase eta, Protein-tyrosine phosphatase receptor type J, PTPRJ, receptor-type tyrosine-protein phosphatase eta, R-PTP-eta, R-PTP-J, SCC1, susceptibility to colon cancer 1, mouse, homolog of | |
Goat | |
Affinity Purified | |
RUO | |
5795 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction