Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat HIF-1 alpha/HIF1A Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 24 publications
Supplier: R&D Systems AF1935
This item is not returnable.
View return policy
Description
HIF-1 alpha Polyclonal specifically detects HIF-1 alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP), Immunocytochemistry, Knockout Validated.Specifications
HIF-1 alpha | |
Polyclonal | |
Unconjugated | |
Q16665.1 | |
HIF1A | |
E. coli-derived recombinant human HIF-1 alpha Arg575-Asn826 Accession # Q16665.1 | |
100 μg | |
Primary | |
Detects human, mouse and rat HIF-1 alpha . | |
Human, Mouse, Rat | |
IgG |
ChIP Assay, Immunocytochemistry, Immunoprecipitation, KnockDown | |
LYOPH | |
Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunoprecipitation 2 ug/500 ug cell lysate, Chromatin Immunoprecipitation (ChIP) 5 ug/5 x 10^6 cells, Immunocytochemistry 0.3-15 ug/mL, Knockout Validated 0.3-15 ug/mL | |
AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF 1A, HIF1, HIF1 alpha, HIF-1 alpha, HIF-1a, HIF1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hypoxia inducible factor 1 alpha subunit, hypoxia inducible factor 1 subunit alpha, hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor), hypoxia-inducible factor 1-alpha, Member of PAS protein 1, member of PAS superfamily 1, MOP1, PAS domain-containing protein 8, PASD8 | |
Goat | |
Affinity Purified | |
RUO | |
3091 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction