Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Neurexophilin-3/NXPH-3 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF5098SP
This item is not returnable.
View return policy
Description
Neurexophilin-3 Polyclonal specifically detects Neurexophilin-3 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Neurexophilin-3 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
KIAA1159, neurexophilin 3, Neurexophilin3, NPH3, NPH3neurexophilin-3, NXPH3 | |
Goat | |
Affinity Purified | |
RUO | |
11248 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
AAI66603 | |
NXPH3 | |
Mouse myeloma cell line NS0-derived recombinant rat Neurexophilin-3/NXPH-3 Gln23-Gly252 Accession # AAI66603 | |
25 μg | |
Primary | |
Detects human, mouse, and rat Neurexophilin-3/NXPH-3 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human Neurexophilin-1/NXPH1 and recombinant rat Neurexophilin-1/NXPH1 is observed. | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction