Learn More
Human/Mouse/Rat TAAR6, Synthetic Peptide, Invitrogen™
Synthetic Peptide
Manufacturer:  Invitrogen GST00005P
Catalog No. PIGST00005P
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Human, Mouse, Rat TAAR6 Synthetic Peptide, GST00005P, from Invitrogen.
GST00005P is the immunizing peptide for the rabbit antibody to human, mouse & rat trace amine-associated receptor 6 (OSR00082W).GST00005P may be used as a control in various immunology applications.Reconstitute with PBS or distilled water.
Specifications
Specifications
Human, Mouse, Rat
Synthetic Peptide
Human, Mouse, Rat
Control
peptide with no preservative
RUO
No preservative
Human/Mouse/Rat TAAR6
Q5QD13, Q923Y5, Q96RI8
Peptide
215855, 294124, 319100
purified
50μg
Lyophilized
Y
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Trace amine-associated receptor 6, Trace amine receptor 4, TaR-4, TA4, TAR4, TRAR4, Ta4, Tar4, Trar4
TAAR6
Control