Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Vinculin Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 8 publications
Supplier: R&D Systems MAB6896
This item is not returnable.
View return policy
Description
Vinculin Monoclonal specifically detects Vinculin in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.Specifications
Vinculin | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 8-25 ug/mL | |
P18206 | |
VCL | |
E. coli-derived recombinant human Vinculin Lys1020-Gln1134 Accession # P18206 | |
100 μg | |
Primary | |
Detects human Vinculin in direct ELISAs and Western blots. | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
728526 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CMD1W, CMH15, Metavinculin, MVCL, VCL, vinculin | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
7414 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction