Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse RBP4/Retinol-Binding Protein 4 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB34762SP
This item is not returnable.
View return policy
Description
RBP4/Retinol-Binding Protein 4 Monoclonal specifically detects RBP4/Retinol-Binding Protein 4 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
RBP4/Retinol-Binding Protein 4 | |
Monoclonal | |
LYOPH | |
Western Blot 0.1-1 ug/mL, Immunoprecipitation 25 ug/mL | |
Q00724 | |
RBP4 | |
Mouse myeloma cell line NS0-derived recombinant mouse RBP4/Retinol-Binding Protein 4 Glu19-Leu201 Accession # Q00724 | |
25 μg | |
Primary | |
Detects mouse RBP4/Retinol-Binding Protein 4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant human RBP4 is observed. | |
Human, Mouse | |
Purified |
Western Blot, Immunoprecipitation | |
423614 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
interstitial, Plasma retinol-binding protein, retinol binding protein 4, plasma, RetinolBinding Protein 4, retinol-binding protein 4, plasma | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5950 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction